Learn More
Contec™ EasyReach™ Cleaning Pads

EasyReach™ Cleaning Pads are very absorbent, which ensures even coverage of disinfectant or IPA.
Brand: Contec™ PSME0001-BPR
380.65 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Single-use sealed polyester
- Provide a low level of particulates which is essential for use in product contact areas
- Very absorbent, ensuring good even coverage of disinfectant or IPA
- Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame
- Sterile cleaning pads are presaturated with 70% IPA

Specifications
Pre-saturated Cleaning Pad | |
19 cm | |
7 cm | |
Not Autoclavable | |
7.5 x 2.8 in. (19 x 7 cm) | |
8 Bags/Cs. |
7.5 in. | |
2.7 in. | |
Quilted Polyester | |
Single-use | |
Rigid, Flexible Film, Half Suits (CAI, CACI), Restricted Access Barrier Systems (RABS), Filling Lines, Laminar Air Flow Hoods (LAF), Bio-Safety Cabinets (BSC), Glove Boxes, Incubators | |
Sterile |
Safety and Handling
- X64 EasyReach Pads presat w/ 70% IPA/30% DI, 2.75" x 7.75", quilted polyester, sterile. BPR
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use dry chemical, carbondioxide for small fires. Use water forlarge fires. to extinguish.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.