Learn More
Contec™ SATWIPES™ Presaturated Wipes



Prepare for a variety of cleanrooms. The SATWIPES™Presaturated Wipes are offered in various options to help tackle general-purpose cleaning in ISO Class 5 to ISO Class 8 environments.
£28.60 - £460.00
Specifications
Product Type | Pre-saturated Wipe |
---|---|
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Sterility | Non-sterile |
Product Code | Brand | Dimensions (L x W) | Material | Saturation Solution | Packaging Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Material | Saturation Solution | Packaging Quantity | Price | Quantity & Availability | ||||||
17294533
![]() |
Contec™
SAT-C1-7030/18-BPR |
23 x 15 cm (9 x 6 in.) | Polyester Cellulose | 70% IPA, 30% DI Water | 100 Pk., 18 Pk./Cs. | N/A | N/A | N/A | |||||
17813294
![]() |
Contec™
SAT-C1-1090 |
15 x 23 cm (6 x 9 in.) | Polyester Cellulose | 10% IPA, 90% DI Water | 12 Pk. |
£306.84
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17284533
![]() |
Contec™
SAT-C1-7030-BPR |
23 x 15 cm (9 x 6 in.) | Polyester Cellulose | 70% IPA, 30% DI Water | 100 Pk., 12 Pk./Cs. |
£375.00
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17214543
![]() |
Contec™
SAT-C3-7030/18-BPR |
23 x 15 cm (9 x 6 in.) | Polyester | 70% IPA, 30% DI Water | 100 Pk., 18 Pk./Cs. |
£458.00
Pack of 1800 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17204543
![]() |
Contec™
SAT-C3-7030-BPR |
23 x 15 cm (9 x 6 in.) | Polyester | 70% IPA, 30% DI Water | 100 Pk., 12 Pk./Cs. |
£460.00
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17224543
![]() |
Contec™
SWNW0013-BPR |
23 x 18 cm (9 x 7 in.) | Polyester, Rayon | 70% IPA, 30% DI Water | 700 Pk., 2 Pk./Cs. |
£159.00
Pack of 1400 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15597979
![]() ![]() |
Contec™
SAT-C1-100 |
23 x 15 cm (9 x 6 in.) | Polyester Cellulose | 100% IPA | 12 Pk. |
£377.00
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
16605544
![]() ![]() |
Contec™
SAT-C3-7030 |
15 x 23 cm (6 x 9 in.) | Polyester | 70% IPA, 30% DI Water | 12 Pk. | N/A | N/A | N/A | |||||
15915760
![]() |
Contec™
SAT-C3-7030/18 |
15 x 23 cm (6 x 9 in.) | Polyester | 750 mL of 70% IPA, 30% DI Water | 1800/Cs. | N/A | N/A | N/A | |||||
17863067
![]() |
Contec™
SAT-C3-AC |
15 x 23 cm (6 x 9 in.) | Polyester | 100% Acetone | 1200/Cs. |
£28.60
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15754047
![]() ![]() |
Contec™
SAT-C3-100 |
15 x 23 cm (6 x 9 in.) | Polyester | 100% IPA | 12 Pk. |
£349.00
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15147915
![]() ![]() |
Contec™
SWNW0013 |
23 x 18 cm (9 x 7 in.) | Polypropylene, Rayon | 70% IPA, 30% DI Water | 700/Bucket | N/A | N/A | N/A | |||||
Description
- Durable, presaturated wipes with a choice of solvent saturations on perforated rolls
- Polyester Nonwoven are ideal for critical applications in ISO Class 5 environments
- SATWIPES with Polyester/Cellulose Nonwoven Wipes offer strength and low extractables for use in less critical applications in ISO Class 6 to 8 environments
- Designed for general-purpose cleaning and surface preparation and for wiping process tools and other equipment and for use prior to cleanroom entry
Order Info
Packaged one hundred 6 x 9 in. (15 x 23cm) wipers on a perforated roll. Most SATWipes cases contain 12 presaturated rolls plus one canister; 19-150-0711 and -0712 packed 18 refills per case without a canister. Custom-blend substrates and additional wiper sizes available through your Fisher Safety Customer Service Representative. Empty canisters for wipes is available: 19-171-955.
Specifications
Pre-saturated Wipe | |
Not Autoclavable | |
Single-use | |
Non-sterile |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.