Learn More
Contec™ VertiKlean™ MAX™ Mop Heads
GREENER_CHOICEBIOPHARMA
Clean walls, ceilings and other hard-to-reach surfaces with ease. VertiKlean™ MAX™ Mop Heads easily snap onto frames and last longer than traditional mops.
Brand:  Contec™ VKSE0300
315.84 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16426830
  • £352.00 / Pack of 20
Estimated Shipment: 15-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features a single layer of hydrophilic polyurethane foam laminated to textured polyester microfiber fabric
  • Backed by a semi-rigid thermoformed plate that easily snaps onto mop head frame
  • Designed for applying disinfectants to both horizontal and vertical surfaces
  • Mop heads are composed of a disposable absorbent sponge laminated to filament polyester knit fabric and backed with a thermoformed plate that easily snaps onto VertiKlean mop hardware
  • Now available with ribbed microfiber for better contamination removal
  • Pivoting mop head frame is ideal for cleaning walls, floors, and ceilings as well as hard-to-reach areas such as fume hoods, mini environments, and around light fixtures and heavy equipment
  • Sterile mop heads are validated sterile per AAMI ISO11137 guidelines
  • Autoclaving mop heads is not recommended due to thermoformed plate; however, all hardware is autoclavable
Order Info
Frames, handles and bucket wringer system sold separately.
Certifications
Validated sterile to a Sterility Assurance Level of 10-6 per AAMI ISO11137:2006 Guidelines. Certificate of sterility is available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
46 x 13 x 1 cm (18 x 5 x 0.375 in.)
White
Not Autoclavable
4/bag; 5 bags/case
Wall Washing System
Knitted Polyester Microfiber, Hydrophilic Urethane Foam
Mop Head
Non-sterile
Single-use
VertiKlean MAX Ribbed Microfiber Mop Head, Sealed Edge
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Contec™ VertiKlean™ MAX™ Mop Heads > Description: VertiKlean MAX Ribbed Microfiber Mop Head, Sealed Edge, Dimensions (L x W x H): 46 x 13 x 1 cm (18 x 5 x 0.375 in.), Sterile: No