Learn More
Decon™ Contrad 70 Liquid Detergent
Concentrated liquid detergent. Ideal for laboratory washing by hand, for soaking or for use with an ultrasonic bath. For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, ferrous metal, ceramic and plastics.
Brand: Decon™ CON705
Description
Specifications
Liquid | |
5 L |
Safety and Handling
- Contrad 70
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305-IF IN EYES:Separate eyelids with finger tips..
- P351-Rinse cautiously with water for several minutes.
- P361-Take off immediately all contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : Potassium hydroxide,Trisodium citrate dihydrate