Learn More
Fisherbrand™ Cool-Pack and Platform
Accessory for use with Horizontal Sub-Gel Electrophoresis Systems.
Brand:  Fisherbrand™ MSMAXICP
 View more versions of this product
Product Code. 11887573
  • £82.80 / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Horizontal Sub-Gel Electrophoresis Systems
Specifications
Specifications
Sub-Gel Maxi Electrophoresis Unit
Cool Pack and Platform
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Cool-Pack and Platform >