missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolBox™ XT Systems
BIOPHARMA

Provide sample cooling or freezing without ice, electricity, or batteries.

Brand:  Corning™ 432026

812.19 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15512801

  • £855.00 / Each
Estimated Shipment: 17-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning CoolBox 2XT systems are bench top cooling workstations that accomodate two CoolRacks and provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.

  • Includes two (2) XT Cooling Cores for 0.5° to 4°C cooling
  • Accommodates two (2) sample modules
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Benchtop cooling system
Cool Box
30 Place
CoolBox 2XT
Green
Enables Cooling for 16 Hours, Freezing for 8 Hours
Holder and Two Cooling Cores
5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number26218018 Certificate TypeCertificate of Analysis Date12/05/2022 Catalog Number432026
Lot Number26218018 Certificate TypeCertificate of Compliance Date28/02/2022 Catalog Number432026
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ CoolBox™ XT Systems > CoolBox 2XT; Holder and two Cooling Cores; Green

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.