missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolCell™ 5mL Filler Vials for CoolCell LX Container
BIOPHARMA

Suitable for repeated use. Corning™ CoolCell™ 5mL Filler Vials ensure cell freezing rate consistency and uniform results when using CoolCell containers.

Brand:  Corning™ 432077

25.03 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 15505535

  • £26.35 / Pack of 6
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.

Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:

  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
5 mL
6
CoolCell 5mL LX Containers
Clear
Glycerol
6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ CoolCell™ 5mL Filler Vials for CoolCell LX Container > 5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.