Learn More
Corning™ CoolCell™ LX Cell Freezing Vial Containers

Provides alcohol-free freezing at the rate of -1°C/minute when placed in a -80°C freezer.
£174.00 - £664.47
Specifications
Material | Polyethylene Foam |
---|---|
Shape | Round |
Type | Freezer Box |
For Use With (Application) | Cryopreservation of Most Cells and Cell Lines |
Holds | 12 (Standard 1 to 2 mL Cryogenic Vials) |
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | |||||
15552771
![]() |
Corning™
432001 |
Purple | 1/Pk., 1/Cs. |
£195.62
Each |
In Stock
Log in to see stock. ![]() |
|||||
15562771
![]() |
Corning™
432002 |
Green | 1/Pk., 1/Cs. |
£191.46
Each |
In Stock
Log in to see stock. ![]() |
|||||
15572771
![]() |
Corning™
432003 |
Orange | 1/Pk., 1/Cs. |
£174.00
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
15582771
![]() |
Corning™
432004 |
Pink | 1/Pk., 1/Cs. |
£196.56
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||
15813885
![]() |
Corning™
432138 |
Assorted (Green, Orange, Pink, Purple) | 4/Pk., 1/Cs. |
£664.47
Pack of 4 |
In Stock
Log in to see stock. ![]() |
|||||
Description
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
- Ease of use
- Alcohol and fluid-free freezing
- Lower cost of use than alcohol-based devices
- High cell recovery and cell viability
- Reproducibility
- Simple, consistent way to standardize controlled-rate freezing
Specifications
Polyethylene Foam | |
Freezer Box | |
12 (Standard 1 to 2 mL Cryogenic Vials) | |
11.7 cm | |
12.7 mm |
Round | |
Cryopreservation of Most Cells and Cell Lines | |
11.7 x 10.11 cm | |
10.11 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.