missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolCell™ LX Cell Freezing Vial Containers
BIOPHARMA

Provides alcohol-free freezing at the rate of -1°C/minute when placed in a -80°C freezer.

Brand:  Corning™ 432004

173.05 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15582771

  • £196.56 / Each
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.

Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:

  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pink
Round
Cryopreservation of Most Cells and Cell Lines
1/Pk., 1/Cs.
11.7 cm
12.7 mm
Polyethylene Foam
Freezer Box
12 (Standard 1 to 2 mL Cryogenic Vials)
11.7 x 10.11 cm
10.11 cm
0.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number26218005 Certificate TypeCertificate of Quality Date17/04/2025 Catalog Number432004
Lot Number26218005 Certificate TypeCertificate of Compliance Date01/07/2023 Catalog Number432004
Lot Number26218005 Certificate TypeCertificate of Analysis Date17/05/2022 Catalog Number432004
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119

    Expires2025-04-30

  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081

    Expires2025-12-31

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    12

    Hours

    3

    Minutes

    23

    Seconds

    25
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ CoolCell™ LX Cell Freezing Vial Containers > 12 x 1 or 2 mL Cryogenic Vials; Pink

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.