Learn More
Eppendorf™ Centrifuge 5920 R
The refrigerated Eppendorf Centrifuge 5920 R offers high-throughput performance and advanced temperature management in a very compact design. With a capacity of up to 4 x 1,000 mL / 52 x 50 mL conical tubes, it can spin both plates and tubes in the same bucket for real flexibility.
£11355.00 - £15690.00
Specifications
Certifications/Compliance | UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers |
---|---|
Max. RCF | 21,194 x g |
Refrigerated | Yes |
Display | Large, Brightly Lit LCD |
Safety Features | Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition |
Product Code | Brand | Model | Rotor | Includes | Electrical Requirements | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Rotor | Includes | Electrical Requirements | Price | Quantity & Availability | ||
15793305
|
Eppendorf™
5948000069 |
Centrifuge 5920 R (EU-IVD), Keypad | None | Without Rotor | 230 V, 50/60 Hz (GB) |
£11910.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16594144
|
Eppendorf™
5948000965 |
Centrifuge 5920 R (EU-IVD), Keypad | S-4 xUniversal-Large | With Rotor S-4 xUniversal-Large, universal Buckets and adapters for 5 mL/15 mL/25 mL/50 mL conical tubes and plates | 230 V, 50/60 Hz (GB) |
£15690.00
1 set |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15753315
|
Eppendorf™
5948000565 |
Centrifuge 5920 R (EU-IVD), Keypad | S-4x750 | With Rotor S-4 x 750, round Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£14755.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15703315
|
Eppendorf™
5948000263 |
Centrifuge 5920 R (EU-IVD), Keypad | S-4x1000 | With Rotor S-4 x 1000, high-capacity Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£15005.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15733315
|
Eppendorf™
5948000468 |
Centrifuge 5920 R (EU-IVD), Keypad | S-4x1000 | With Rotor S-4 x 1000, plate/tube Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£14515.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15773315
|
Eppendorf™
5948000662 |
Centrifuge 5920 R (EU-IVD), Keypad | S-4x1000 | With Rotor S-4 x 1000, plate/tube Buckets and adapters for round-bottom tubes with diameter 13/16 mm | 230 V, 50/60 Hz (GB) |
£14515.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15723315
|
Eppendorf™
5948000360 |
Centrifuge 5920 R (EU-IVD), Keypad | S-4x1000 | With Rotor S-4 x 1000, round Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£14515.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15895123
|
Eppendorf™
5948000060 |
Centrifuge 5920 R, Keypad | None | Without Rotor | 230 V, 50/60 Hz (GB) |
£11355.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16584144
|
Eppendorf™
5948000961 |
Centrifuge 5920 R, Keypad | S-4 xUniversal-Large | With Rotor S-4 xUniversal-Large, universal Buckets and adapters for 5 mL/15 mL/25 mL/50 mL conical tubes and plates | 230 V, 50/60 Hz (GB) |
£14995.00
1 set |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15825143
|
Eppendorf™
5948000860 |
Centrifuge 5920 R, Keypad | S-4 xUniversal-Large | With Rotor S-4 xUniversal-Large, universal Buckets and adapters for round-bottom tubes with diameter 13/16 mm | 230 V, 50/60 Hz (GB) |
£14630.00
1 set |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15875133
|
Eppendorf™
5948000560 |
Centrifuge 5920 R, Keypad | S-4x750 | With Rotor S-4 x 750, round Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£14060.00
1 set |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15815133
|
Eppendorf™
5948000260 |
Centrifuge 5920 R, Keypad | S-4x1000 | With Rotor S-4 x 1000, high-capacity Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£14295.00
1 set |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16662842
|
Eppendorf™
5948000182 |
Centrifuge 5920 R, Keypad | S-4x1000 | With Rotor S-4x1000, plate/tube Bucket and plate carrier | 230 V, 50/60 Hz (GB) |
£14070.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15855133
|
Eppendorf™
5948000460 |
Centrifuge 5920 R, Keypad | S-4x1000 | With Rotor S-4 x 1000, plate/tube Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£13810.00
1 set |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15835133
|
Eppendorf™
5948000365 |
Centrifuge 5920 R, Keypad | S-4x1000 | With Rotor S-4 x 1000, round Buckets and adapters for 15 mL/50 mL conical tubes | 230 V, 50/60 Hz (GB) |
£13810.00
1 set |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
cooled, with rotor S-4xUniversal-Large, incl. Universal beakers and adapters for conical tubes 5 mL / 15 mL / 25 mL / 50 mL and plates, 230 V / 50 - 60 Hz
- Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 1,000 mL
- Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
- Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
- High centrifugation speed of up to 21,194 x g (13,700 rpm)
- Aerosol-tight Eppendorf QuickLock™ caps and lids allow fast and ergonomic lid locking
- Centrifuge lid with soft-touch lid closure
- Quiet operation to improve your work environment
- Compact footprint saves valuable bench space
- Automatic rotor recognition and imbalance detection for maximum operational safety
Specifications
UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers | |
Yes | |
Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition | |
Benchtop Centrifuge | |
74 cm | |
40 cm | |
1650 W |
21,194 x g | |
Large, Brightly Lit LCD | |
10 sec. to 99 hr. 59 min., Continuous Run Function, Short-spin | |
29.2 in. | |
15.9 in. | |
230 V |
The Centrifuge 5920 R is designed for separating liquid substance mixtures with different densities, in particular, for processing and analyzing samples from the human body in in-vitro diagnostic applications to ensure that the in-vitro diagnostic device can be used according to its intended purpose. This centrifuge including components is an in-vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.