Learn More
Corning™ CoolRack™ CF Cryogenic Vial and FACS Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Brand: Corning™ 432049
This item is not returnable.
View return policy
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.Specifications
Capacity: 15 cryogenic vials or FACS tubes | |
CF15 | |
Cryogenic Vial, FACS Tube Module |