missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ CF Cryogenic Vial and FACS Tube Modules
BIOPHARMA

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

£167.00 - £344.00

Specifications

Type Cryogenic Vial, FACS Tube Module
View More Specs

Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15542821
BIOPHARMA
View Documents
Corning™
432049
Capacity: 15 cryogenic vials or FACS tubes
£167.00
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
15562821
BIOPHARMA
View Documents
Corning™
432051
Capacity: 45 cryogenic vials or FACS tubes
£311.00
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
15572821
BIOPHARMA
View Documents
Corning™
432052
Capacity: 30 cryogenic vials or FACS tubes, with gripping wells for one-hand 1 vial opening/closing
£344.00
Each
Estimated Shipment: 31-12-2024
to see stock.
Add to basket
 
Description

Description

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • Ensure +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Specifications

    Specifications

    Cryogenic Vial, FACS Tube Module
    Videos
    SDS
    Documents

    Documents

    Product Certifications