Learn More
Corning™ CoolRack™ M Microcentrifuge Tube Modules
Thermo-conductive microfuge tube modules that eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources.
Brand: Corning™
Code : NEW
Additional Details : Weight : 5.00000kg
Description
Corning CoolRack thermo-conductive microcentrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
Specifications
Gray | |
Microcentrifuge Tubes | |
6 x 15 | |
Microcentrifuge Tube Modules |
Cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen | |
90 x 1.5 or 2 mL Microcentrifuge Tubes | |
M90 |