Learn More
Corning™ CoolRack™ M Microcentrifuge Tube Modules
Thermo-conductive microfuge tube modules that eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources.
£65.60 - £560.00
Specifications
For Use With (Application) | Cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen |
---|---|
For Use With (Equipment) | Microcentrifuge Tubes |
Type | Microcentrifuge Tube Modules |
Product Code | Brand | Holds | Color | Design | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Color | Design | Price | Quantity & Availability | |||
15592801
|
Corning™
432034 |
6 x 1.5 or 2 mL Microcentrifuge Tubes | Gray | M6 |
£65.60
Each |
In Stock
Log in to see stock. |
|||
15502811
|
Corning™
432035 |
6 x 1.5 or 2 mL Microcentrifuge Tubes | Green | M6 |
£65.60
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15512811
|
Corning™
432036 |
6 x 1.5 or 2 mL Microcentrifuge Tubes | Orange | M6 |
£65.60
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15522811
|
Corning™
432037 |
15 x 1.5 or 2 mL microcentrifuge tubes | Gray | M15 |
£159.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15532811
|
Corning™
432038 |
15 x 1.5 or 2 mL microcentrifuge tubes | Green | M15 |
£159.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15542811
|
Corning™
432039 |
15 x 1.5 or 2 mL microcentrifuge tubes | Orange | M15 |
£164.09
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15552811
|
Corning™
432040 |
24 x 1.5 or 2 mL microcentrifuge tubes; SBS-compatible | Gray | M24 |
£278.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15562811
|
Corning™
432041 |
30 x 1.5 or 2 mL Microcentrifuge Tubes | Gray | M30 |
£288.96
Each |
In Stock
Log in to see stock. |
|||
15572811
|
Corning™
432042 |
30 x 1.5 or 2 mL Microcentrifuge Tubes | Green | M30 |
£280.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15582811
|
Corning™
432043 |
30 x 1.5 or 2 mL Microcentrifuge Tubes | Orange | M30 |
£280.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15592811
|
Corning™
432044 |
90 x 1.5 or 2 mL Microcentrifuge Tubes | Gray | M90 |
£421.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15502821
|
Corning™
432045 |
96 x 1.5 or 2 mL Microcentrifuge Tubes with A to H and 1 to 12 | Gray | M96 |
£560.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
Description
Corning CoolRack thermo-conductive microcentrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
Specifications
Cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen | |
Microcentrifuge Tubes | |
Microcentrifuge Tube Modules |