Learn More
Copper(II) bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
£17.70 - £471.00
Chemical Identifiers
CAS | 7789-45-9 |
---|---|
Molecular Formula | Br2Cu |
Molecular Weight (g/mol) | 223.36 |
MDL Number | MFCD00010970 |
Synonym | Cupric bromide |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10422401
|
Thermo Scientific Acros
206330050 |
5 g | Glass bottle |
£17.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144880
|
Thermo Scientific Acros
206331000 |
100 g | Glass bottle |
£35.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10398653
|
Thermo Scientific Acros
206335000 |
500 g | Glass bottle |
£117.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10493065
|
Thermo Scientific Acros
206330025 |
2.5 kg | Glass bottle |
£471.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7789-45-9 | |
223.36 | |
Cupric bromide |
Br2Cu | |
MFCD00010970 |
Specifications
900.0°C | |
7789-45-9 | |
Crystalline Powder | |
Br2Cu | |
MFCD00010970 | |
15, 2619 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4 | |
223.36 | |
Extra Pure | |
Copper(II) bromide |
498.0°C | |
Black-Gray to Gray | |
99% min. (Iodometry) | |
CuBr2 | |
01,161; 02,84; 05,158; 06,138; 13,112; 15,100 | |
Cupric bromide | |
223.36 | |
≥99% | |
4.77 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-167-2
missing translation for 'tsca' : TSCA
- Copper(II) bromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only