missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) sulfate, anhydrous, Reagent Grade, Thermo Scientific Chemicals Product Code.: 11357628

Copper(II) sulfate, anhydrous, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11357628
500 g
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11357628

Brand: Thermo Scientific Alfa Aesar 033308.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in the Fehling's solution and Benedict's solution to test for reducing sugars. Also, it is used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagleShow MoreShow Less
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Melting Point >560°C (decomposition)
Color Gray
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
Formula Weight 159.6
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Density 3.6
Chemical Name or Material Copper(II) sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW19J012 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number033308.A3,033308.36,033308.22
Lot NumberP17J032 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number033308.36
Lot NumberR03I033 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number033308.A3,033308.36,033308.22
Lot NumberE10J18 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number033308.A3,033308.36,033308.22
Lot NumberF11S037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033308.A3,033308.36,033308.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) sulfate, anhydrous, Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.