missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) sulfate, anhydrous, Reagent Grade, Thermo Scientific Chemicals Product Code.: 11377628

Copper(II) sulfate, anhydrous, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11377628
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11377628

Brand: Thermo Scientific Alfa Aesar 033308.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in the Fehling's solution and Benedict's solution to test for reducing sugars. Also, it is used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagleShow MoreShow Less
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Melting Point >560°C (decomposition)
Color Gray
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
Formula Weight 159.6
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Density 3.6
Chemical Name or Material Copper(II) sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW19J012 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number033308.A3,033308.36,033308.22
Lot NumberR03I033 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number033308.A3,033308.36,033308.22
Lot NumberE10J18 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number033308.A3,033308.36,033308.22
Lot NumberF11S037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033308.A3,033308.36,033308.22
Lot NumberB19T023 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033308.A3,033308.36,033308.22
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Copper(II) sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) sulfate, anhydrous, Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.