missing translation for 'onlineSavingsMsg'
Learn More
Copper(Ii) Sulfate, Honeywell Fluka™ Product Code.: 15641220

Copper(Ii) Sulfate, Honeywell Fluka™
BIOPHARMA

Product Code. 15641220 Shop All Honeywell Fluka Products
1 kg
Click to view available options
Quantity:
250 g
1 kg
6 x 250 g
50 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15641220

Brand: Honeywell-Fluka 128521KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., anhydrous, meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula CuSO4
UN Number 3077
Formula Weight 159.61g/mol
Percent Purity 99 to 100.5% (based on anhydrous substance)
Grade puriss.
Vapor Pressure 7.3mmHg (25°C)
Density 3.603g/mL at 25°C
Chemical Name or Material Copper(Ii) Sulfate, Meets analytical specification of Ph. Eur.BP, USP

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0430143 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G
Lot NumberA0404624 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G
Lot NumberA0444906 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G
Lot NumberA0432945 Certificate TypeCertificate of Analysis Date09/08/2021 Catalog Number12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G
Lot NumberA0396738 Certificate TypeCertificate of Analysis Date30/11/2020 Catalog Number12852-1KG,128520250,128521000,12852-6X1KG,12852-6X250G,12852-250G
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Copper sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(Ii) Sulfate, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.