missing translation for 'onlineSavingsMsg'
Learn More

Copper, powder, 99+%, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar A16243.0E

162.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11319236

  • £162.81 / 2500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7440-50-8
2595°C
2500 g
1083°C
Cu
≥99%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253412 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberA16243.22,A16243.36,A16243.0E
Lot Number10239201 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16243.22,A16243.36,A16243.0E
Lot Number10222464 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16243.22,A16243.36,A16243.0E
Lot Number10183791 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16243.22,A16243.36,A16243.0E
Lot Number10054504 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16243.22,A16243.36,A16243.0E
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper, powder, 99+%, Thermo Scientific Chemicals > 2500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.