missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Untreated 245mm Square BioAssay Dishes
Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.
£203.35 - £250.79
Specifications
Closure Color | Clear |
---|---|
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Square |
Capacity (Metric) | 150 mL (Well) |
Product Code | Brand | Dimensions (L x W) | Depth (Metric) Inside | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Depth (Metric) Inside | Description | Price | Quantity & Availability | |||
10002611
|
Corning™
431272 |
245 x 245 mm | 18 mm | With handles |
£203.35
Pack of 16 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10643053
|
Corning™
431301 |
245 x 245 mm | 12.5 mm | Without handles |
£250.79
Pack of 20 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10737041
|
Corning™
431111 |
245 x 245 mm | 18 mm | Without handles |
£203.35
Pack of 16 |
In Stock
Log in to see stock. |
|||
Description
Improve the efficiency of microbial assays including library replication, colony picking, and plaque lift studies.
- Polystyrene dishes feature stacking beads for stable stacking
- Compatible with various automated colony picking and gridding robots
- Supplied with vented lid to provide consistent gas exchange
- Sterile, certified nonpyrogenic
- Patented (U.S. Patent No.. 5,856,176) design allows for easier handling
Specifications
Clear | |
Polystyrene | |
150 mL (Well) | |
500 sq. cm | |
Polystyrene | |
Sterile | |
Lid | |
245 mm | |
25 mm |
Closure Included | |
Square | |
Clear | |
Single-use | |
Square | |
Cell Culture Dish | |
245 x 245 mm | |
245 mm |