Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Brand: Corning™ 3577
Code : NEW
Additional Details : Weight : 5.12559kg
Description
- Minimal well to well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 112μL
- Working volume: 20μL to 80μL
- Surface area/well: 0.06cm2
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Plate dimensions meet proposed industry standards
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
High Binding | |
Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions. Finding capacity is approximately 400 to 500ng lgG/cm2 |
Without Lid | |
Black | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. |