missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
£363.97 - £682.93
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Material | Polystyrene |
No. of Wells | 384 |
Sterility | Non-sterile |
Product Code | Brand | Color | Well Shape | Surface Treatment | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Well Shape | Surface Treatment | Price | Quantity & Availability | |||
10713767
|
Corning™
3577 |
Black | Flat Bottom (F-well) | High Binding |
£472.51
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10382883
|
Corning™
3574 |
White | Flat Bottom (F-well) | Non-binding |
£582.50
Pack of 50 |
In Stock
Log in to see stock. |
|||
10087842
|
Corning™
3575 |
Black | Flat Bottom (F-well) | Non-binding |
£576.32
Pack of 50 |
In Stock
Log in to see stock. |
|||
10745303
|
Corning™
3573 |
Black | Flat Bottom (F-well) | Non-treated |
£363.97
Pack of 50 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||
15328271
|
Corning™
4510 |
Black | Round Bottom (U-well) | High Binding |
£573.17
Pack of 50 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||
15327190
|
Corning™
4512 |
White | Round Bottom (U-well) | Non-treated |
£518.34
Pack of 50 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||
15284518
|
Corning™
4511 |
Black | Round Bottom (U-well) | Non-treated |
£463.44
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
15314322
|
Corning™
4514 |
Black | Round Bottom (U-well) | Non-binding |
£637.33
Pack of 50 |
In Stock
Log in to see stock. |
|||
15338271
|
Corning™
4513 |
White | Round Bottom (U-well) | Non-binding |
£682.93
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
- Minimal well to well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 112μL
- Working volume: 20μL to 80μL
- Surface area/well: 0.06cm2
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Plate dimensions meet proposed industry standards
Specifications
Standard | |
Polystyrene | |
Non-sterile |
16 x 24 | |
384 |