missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Nonbinding Surface (NBS™) Microplates
Enhanced signal-to-noise ratio
Brand: Corning™ 3653BC
Code : NEW
Additional Details : Weight : 9.25328kg
Description
- Ideal for homogenous assays such as Scintillation Proximity Assays (SPA) and fluorescent assays
- Modified polymer surface is nonionic hydrophilic, minimizing bio-interaction within wells to reduce nonspecific binding and reagent costs
- Significantly reduces protein and nucleic acid binding to polymers, maintains enzyme activity, and inhibits adhesion of a number of cell lines Stable to aqueous solutions containing EtOH
- Use at temperature range of 4° to 37°C (39° to 99°F)
- Flat-bottom wells
- Compatible with a wide range of plate readers, liquid handlers, and robotic systems
Generic Bar Coded Plates
- Code 128 bar code format ensures compatibility with most bar code scanning and software systems
- To avoid mix-ups, the surface treatment of the microplate wells is easily identified
- Labels applied to all four sides of the plate No sequence number duplicates
Specifications
Includes Barcode | |
16 x 24 | |
Plate readers, liquid handlers and robotic systems | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
39°F to 99°F | |
Clear |
Standard | |
White | |
112 μL | |
20 to 80 μL | |
Non-binding | |
For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays | |
4°C to 37°C |