missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Glass Multiwell Culture Slide
GREENER_CHOICE

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Brand:  Corning™ 354630

 View more versions of this product

Product Code. 10595431

  • £375.00 / Pack of 12
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Culture Slide
    3/Pk.
    Collagen I
    Adhesion, growth and differentiation of neuronal and transfected cell lines
    12/Cs.
    8
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers