Learn More
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
GREENER_CHOICE
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand:  Corning™ 354401
 View more versions of this product
Product Code. 10543051
  • £106.65 / Pack of 20
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Order Info
Bulk quantities available; please contact your Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Polystyrene
Clear
Polystyrene
Collagen Type I
Lid
60 mm
Closure Included
Round
21.3 sq. cm
Round
Cell Culture Dish
20/cs.