Learn More
Corning™ BioCoat™ Poly-D-Lysine

Recommended for the culture of neurons, glial cells, and transfected cells
Brand: Corning™ 354210
Description
Corning™ Poly-D-Lysine, 20mg, is used to culture a wide variety of cell types, particularly neurons, glial cells, and transfected cells.
Source
Synthetic
Quality
Shown to promote cell attachment and spreading of rat cerebellar granule cells
Found negative for bacteria, fungi, and mycoplasma
Molecular Weight
500-550 kD
Preparation and Storage
Stable for three months at 2° to 8°C (lyophilized) or three months at -20°C (solubilized
Handling
Recommended concentration: 2.5-5 μg/cm² of growth surface depending on cell type
Formulation
Lyophilized from aqueous solution; reconstitute in dH20

Specifications
2° to 8°C | |
Lyophilized | |
20 mg |
Recommended concentration: 2.5-5μg/cm2 of growth surface depending on cell type | |
Poly-D-Lysine | |
Synthetic |
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.