Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand:  Corning™ 356467
Code : 67
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 10478822
  • £420.44 / Pack of 100
Estimated Shipment: 03-12-2024
to see stock.
Add to basket
Description
Description
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Coated with 0.2 micron filtered PDL in a cleanroom
Note: Dishes are not terminally sterilized and not assigned an SAL level. 
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
Clear
Polystyrene
Clear
Polystyrene
Poly-D-Lysine
Lid
35 mm
Closure Included
Round
9.6 sq. cm
Round
Cell Culture Dish
100/cs.