Learn More
Corning™ Black and White 96-Well Polypropylene Assay Plates
Lower compound sticking and higher solvent resistance than polystyrene plates
Brand: Corning™ 3355
Additional Details : Weight : 4.74000kg
Description
- Ideal for fluorescence and luminescence assays and for compound storage
- Round-bottom wells
- Untreated surface
Specifications
Standard | |
White | |
96 | |
Non-sterile | |
Round |
8 x 12 | |
Polypropylene | |
75 to 200 μL | |
Non-treated | |
Ideal for fluorescence and luminescence assays and for compound storage |