missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Ampicillin, Sodium Salt

Packaged in plastic bottles

Brand:  Corning™ 61-238-RH

Code : NEW

Additional Details : CAS Number : 56-75-7 Weight : 1.00000kg

 View more versions of this product

Product Code. 15333771

  • £99.65 / 10g
Estimated Shipment: 06-01-2025
to see stock.

Description

Description

Specifications

Specifications

2° to 8°C
Antibiotic
10 g
Powder
Plastic Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers