missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Ampicillin, Sodium Salt

Packaged in plastic bottles

Brand:  Corning™ 61-238-RM

Code : NEW

Additional Details : CAS Number : 86393-32-0 Weight : 0.16200kg


 View more versions of this product

Product Code. 15343771

  • £351.00 / 100g
Estimated Shipment: 06-01-2025
to see stock.

Description

Description

Specifications

Specifications

2° to 8°C
Antibiotic
100 g
Powder
Plastic Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ampicillin sodium salt

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : monosodium ampicillin salt

SDS
Documents

Documents

Certificates
Special Offers

Special Offers