Learn More
Corning™ cellgro™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix

Used for culture of epithelial, endothelial, and granulosa cell types under low serum conditions. Corning™ cellgro™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix has low levels of endotoxin, and absence of contamination.
Brand: Corning™ 10-092-CV
Description
- Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Fifty-fifty mixture of DMEM and F-12, originally formulated for rat neuroblastoma cells and MDCK cells
- Serum-free growth may be possible with appropriate protein supplements
- Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
- Formulation of DMEM/Ham's F-12 contains L-glutamine
- Sterile

Specifications
2° to 8°C | |
Liquid | |
Sterile | |
<0.25EU/mL | |
To assess growth promotion and cytotoxicity characteristics | |
6 x 500 mL |
1 X | |
DMEM and Ham's F-12, 50/50 Mix | |
Glutamine, HEPES | |
Inorganic Salts: 116.65mg/L CaCl2 (anhydrous), 0.417mg/L FeSO4.7H2O, 0.05mg/L Fe(NO3)3.9H2O, 311.8mg/L KCl, 0.0008mg/L CuSO4 (anhydrous), 84.95mg/L MgSO4 (anhydrous), 7000mg/L NaCl, 62.5mg/L Na2HPO4.H2O, 71mg/L Na2HPO4 (anhydrous), 2438mg/L NaHCO3, 0.4315mg/L ZnSO4.7H2O; Amino Acids: 4.45mg/L L-Alanine, 147.5mg/L L-Arginine. HCl, 7.5mg/L L-Asparagine.H2O, 6.65mg/L L-Aspartic acid, 17.56mg/L L-Cysteine.HCl.H2O, 31.285mg/L L-Cysteine.2HCl, 7.35mg/L L-Glutamic acid, 365.1mg/L L-glutamine, 18.75mg/L Glycine, 31.48mg/L L-Histidine.HCl.H2O, 54.37mg/L L-Isoleucine, 58.95mg/L L-Leucine, 91.35mg/L L-Lysine. HCl, 17.24mg/L L-Methionine, 35.48mg/L L-Phenylalanine, 17.25mg/L L-Proline, 26.25mg/L L-Serine, 53.55mg/L L-Threonine, 9.02mg/L L-Tryptophan, 55.815mg/L L-Tyrosine.2Na.2H2O, 52.85mg/L L-Valine; Vitamins: 0.00365mg/L Biotin, 2.24mg/L D-Ca-Pantothenate, 8.98mg/L Choline chloride, 2.65mg/L Folic acid, 12.61mg/L i-Inositol, 2.0185mg/L Nicotinamide, 2.031mg/L Pyridoxine. HCl, 0.219mg/L Riboflavin, 2.17mg/L Thiamine. HCl; Other: 3151mg/L D-Glucose, 2.385mg/L Hypoxanthine, Sodium , 0.105mg/L DL-Thioctic (lipoic) acid, 0.044mg/L Methyl lineoleate, 8.1mg/L Phenol red, Sodium, 0.08mg/L Putrescine.2HCl, 110mg/L Sodium pyruvate, 0.365mg/L Thymidine, 3574.8mg/L HEPES. | |
7 to 7.4 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
28924014 | Certificate of Quality | 21/02/2025 | 10-092-CV |
28924014 | Certificate of Quality | 19/02/2025 | 10-092-CV |
17624014 | Certificate of Quality | 18/12/2024 | 10-092-CV |
09324010 | Certificate of Quality | 05/11/2024 | 10-092-CV |
00224014 | Certificate of Quality | 18/09/2024 | 10-092-CV |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.