missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.

Brand:  Thermo Scientific™ 4116-2000

72.07 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10434162

  • £176.00 / Pack of 4
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Save preparation time and avoid contamination risk by using Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

  • Sterile and single use to reduce contamination risk
  • Collapses when autoclaved to reduce bio hazardous waste
  • Graduated for quick volume assessment
  • Excellent validation support: Validation binder available upon request†

Note:

† Validation binders are available to customers as support documentation during your product validation process. This confidential information can be requested at: rocregsupport@thermofisher.com. Following your request submission, a Confidentiality Agreement will be forwarded to you for completion.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

64 oz.
PETG
1
None
Single-use
Narrow
Baffled
45-430 mm
2000 mL
4
Erlenmeyer
Not Autoclavable
Yes
Sterile
Vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1419796 Certificate TypeCertificate of Processing Date18/02/2025 Catalog Number4116-2000
Lot Number1347999 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4116-2000
Lot Number1334358 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4116-2000
Lot Number1334360 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4116-2000
Lot Number1419796 Certificate TypeCertificate of Compliance Date25/01/2025 Catalog Number4116-2000
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile > Sterile PETG Erlenmeyer Flask; 2000mL; Baffled Bottom; Blue, Vented Closure

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.