Learn More
Corning™ PETG Baffled Erlenmeyer Flask with Vent Cap
Constructed of optically clear polyethylene terephthalate (PETG) and ideal for shaker culture applications.
Brand: Corning™ 431281
Description
The Corning™ Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are ready-to-use, ideal for shaker culture applications, and storage.
- Available with baffled bottom
- Molded-in graduations for accuracy
- Large vent cap for continuous gas exchange while preventing leakage
- Highest Sterility Assurance Level (SAL) of 10−6
- Nonpyrogenic and RNase-/DNase-free
- Made from optically-clear polyethylene terephthalate(PTEG)
Specifications
Nonpyrogenic, DNase/RNase-free | |
Polypropylene | |
6/Cs. | |
Baffled Erlenmeyer | |
Vented |
Orange | |
2 L | |
1/Pk. | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.