Learn More
Corning™ Collection Bags
Collection bags with three ports used for biopharmaceutical applications
Brand:  Corning™ 91-200-45
 View more versions of this product
Product Code. 15796278
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 L
EVA
1/Pk.
Each
Heat sealable flexible tubing
1/Ea.
Port 1 - 0.37 in. Connector, 12 L x 0.37 I.D. x 0.62 in. O.D., chemically resistant, heat sealable flexible tubing, port 2 - male MPC connector with 0.37 in. barb, female MPC cap, port- 3 female MPC connector with 0.37 in. barb, male MPC plug, break-off i