missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Collection Bags

Collection bags with three ports used for biopharmaceutical applications

Brand:  Corning™ 91-200-83

Additional Details : Weight : 0.43600kg

 View more versions of this product

Product Code. 17223584

  • £301.35 / Each
Estimated Shipment: 11-12-2024
to see stock.

Description

Description

Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.

Specifications

Specifications

200 L
Heat sealable flexible tubing
1/Ea.
60 L x 0.5 I.D. x 0.75 in. O.D. and 40 L x 0.12 I.D. x 0.25 in. O.D. chemically resistant, heat sealable flexible tubings, female luer with 0.15 in. barb, swabbable valve, non-breather female luer cap with female MPX cap and male MPX connector with 0.5 in
Biopharmaceutical applications
ULDPE
1/Pk.
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers