missing translation for 'onlineSavingsMsg'
Learn More

Corning™ G418 Sulfate, 50mg/mL
BIOPHARMA

Blocks polypeptide synthesis by hindering protein elongation. Corning™ G418 Sulfate is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.

Brand:  Corning™ 30-234-CR

175.27 GBP valid until 2025-12-31
Use promo code "24361" to get your promotional price.


 View more versions of this product

Product Code. 15303681

  • £241.00 / 20mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Well-known, effective selection agent G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 μg/mL. This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene.  G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
Liquid
Sterile
20 mL
50 mg/ mL Solution (active G418)
Antibiotic
Plastic Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number30234462 Certificate TypeCertificate of Quality Date13/04/2025 Catalog Number30-234-CR
Lot Number30234459 Certificate TypeCertificate of Quality Date28/11/2024 Catalog Number30-234-CR
Lot Number30234457 Certificate TypeCertificate of Quality Date03/09/2024 Catalog Number30-234-CR
Lot Number30234460 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog Number30-234-CR
Lot Number30234452 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number30-234-CR
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ G418 Sulfate, 50mg/mL > G418 Sulfate, Liquid; 50mg/mL Solution (active G418); 20mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.