Learn More
Corning™ Lymphocyte Separation Medium
Designed for the in vitro isolation of lymphocytes from diluted whole blood.
Brand: Corning™ 25-072-CV
Code : NEW
Additional Details : Weight : 0.55800kg
Description
Lymphocyte Separation Medium was originally designed for the in vitro isolation of lymphocytes from diluted whole blood. It is a sterile filtered, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/cm3 at 20°C.
Specifications
15° to 30°C | |
Lymphocyte separation medium | |
1 x 500 mL |
Liquid | |
Sterile |