missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Trypan Blue
BIOPHARMA

Can be used for variety cell counting methods requiring information on cell count and viability.

Brand:  Corning™ 25-900-CI


Product Code. 15393661

  • £39.60 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • 0.4% (w/v) solution
  • In PBS
  • pH 7.5 ±0.5
  • Quantity: 100mL
  • Viable cells exclude the dye, while nonviable cells absorb the dye and appear blue
  • Cells should be in suspension as single cells in buffered saline before counting
  • Can be used in assays providing total cell count, viable cell or nonviable cell count alone
  • Solution is provided at the common concentration of 0.4% (w/v)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Trypan Blue 0.4% Solution in PBS
Liquid
Plastic Bottle
7.5 ±0.5
100 mL
15° to 30°C
Sterile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Trypan Blue Solution

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2

Hazard Statement
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number06524007 Certificate TypeCertificate of Quality Date19/02/2025 Catalog Number25-900-CI
Lot Number06524007 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number25-900-CI
Lot Number28923007 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog Number25-900-CI
Lot Number03723005 Certificate TypeCertificate of Analysis Date30/06/2023 Catalog Number25-900-CI
Lot Number18622017 Certificate TypeCertificate of Analysis Date27/04/2023 Catalog Number25-900-CI
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Trypan Blue > Trypan Blue Solution 0.4%; 1 x 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.