Learn More
Corning™ Clear Polypropylene 384-Well Deep-Well Plates
Ideal for high-throughput applications requiring added volume
Brand:  Corning™ 3342
Code : 66
Additional Details : Weight : 4.49060kg
 View more versions of this product
Product Code. 10214932
  • £378.60 / Pack of 50
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
Specifications
Specifications
Standard
16 x 24
Polypropylene
Sterile
Conical Bottom (V-well)
50/Cs.
50/Cs.
Without Lid
Clear
384
Non-treated
For high throughput applications requiring added volume
5/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products