missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Clear Polypropylene 384-Well Deep-Well Plates
Ideal for high-throughput applications requiring added volume
£298.05 - £378.60
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 16 x 24 |
Color | Clear |
Material | Polypropylene |
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
10412952
|
Corning™
3347 |
Non-sterile |
£298.05
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
10214932
|
Corning™
3342 |
Sterile |
£378.60
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
Order Info
Packaged in bags of ten
Certifications
Certified DNase- and RNase-free
Specifications
Standard | |
16 x 24 | |
Polypropylene | |
Non-treated | |
For high throughput applications requiring added volume | |
5/Pk. |
Without Lid | |
Clear | |
384 | |
Conical Bottom (V-well) | |
50/Cs. | |
50/Cs. |