Learn More
Corning™ Collagen I, Human
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Brand:  Corning™ 354243
 View more versions of this product
Product Code. 11503560
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.2 to 2 μg/cm2
Frozen in 2 mm hydrogen chloride
Human Placenta
-20°C
Cell Attachment and Proliferation, Cell-specific Morphology and Function
0.25 mg
Collagen, Human Type I
>90% by SDS-PAGE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Collagen I, Human > 0.25 mg; Collagen I, Human