missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Collagen I, Rat
GREENER_CHOICEBIOPHARMA

Enhances cell attachment and proliferation or promotes expression of cell-specific morphology and function. Corning™ Collagen I, Rat is effective as a gel or a thin coating at a recommended concentration of 5 to 10μg/cm2 of growth surface depending on cell type.

Brand:  Corning™ 354236

202.35 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 11563550

  • £218.33 / 100mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 to 4 mg/mL
Liquid in 0.02N acetic acid
Rat Tail Tendon
2°C to 8°C
Cell Attachment and Proliferation, Cell-specific Morphology and Function
100 mg
Collagen, Rat Tail Type I
>90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number00925002 Certificate TypeCertificate of Quality Date21/04/2025 Catalog Number354236
Lot Number34024001 Certificate TypeCertificate of Quality Date16/04/2025 Catalog Number354236
Lot Number27724003 Certificate TypeCertificate of Quality Date24/12/2024 Catalog Number354236
Lot Number21424001 Certificate TypeCertificate of Quality Date28/11/2024 Catalog Number354236
Lot Number19324001 Certificate TypeCertificate of Quality Date15/10/2024 Catalog Number354236
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Collagen I, Rat > 100 mg; Collagen I, Rat Tail

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.