missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Untreated Culture Dishes
GREENER_CHOICE

Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.

Brand:  Corning™ 430591

 View more versions of this product

Product Code. 10419141

  • £383.93 / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from optically clear virgin polystyrene
  • Not treated for applications where cell attachment is not desired
  • Not treated for applications where cell attachment is not desired
  • Stacking beads aid in handling
  • Vents provide consistent gas exchange
  • Sterile
  • Sterile
  • Nonpyrogenic


Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.

Cat. no. 431111, Square dishes with interior bottom dimensions of 224 x 224 mm.

Order Info

For Cell Culture Treated Polystyrene Dishes: Shelf pack 430293 consists of six-pack carriers which hold six sleeves of 10 dishes each–minimizing contamination of unused dishes after sleeve is opened. Supplied in sleeves of 20 dishes (except 150 x 25mm dish which is packaged five per sleeve and 245 x 245mm dish which comes four per sleeve).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
16.5 mL (Well)
55 cm2
Polystyrene
Sterile
Cell Culture Petri Dish
500/Cs.
20 mm
Yes
Flat
Clear
Single-use
Round
Non-treated
Lid
20/Pk.
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Untreated Culture Dishes > 100 x 20 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.