Learn More
Corning™ DNA-BIND™ 96-Well Plates
Convenient method for covalently immobilizing ssDNA by either the 5' or 3' end
Brand: Corning™ 2499
This item is not returnable.
View return policy
Description
- Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
- Ideal for hybridization and amplification
- Surface provides binding with high specificity and extremely low background
- Flat-bottom wells
Specifications
Standard | |
White | |
96 | |
DNA-Bind | |
Ideal for hybridization and amplification | |
White |
8 x 12 | |
Polystyrene | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
50/Cs. |