missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DNA-BIND™ 96-Well Plates
Convenient method for covalently immobilizing ssDNA by either the 5' or 3' end
£158.40 - £750.14
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polystyrene |
No. of Wells | 96 |
Sterility | Non-sterile |
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
10525341
|
Corning™
2505 |
Clear | 50/Cs. |
£750.14
Pack of 50 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
10286671
|
Corning™
2525 |
Clear | 10/Cs. | ||||||
10691072
|
Corning™
2499 |
White | 50/Cs. | ||||||
Description
- Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
- Ideal for hybridization and amplification
- Surface provides binding with high specificity and extremely low background
- Flat-bottom wells
Specifications
Standard | |
Polystyrene | |
Non-sterile |
8 x 12 | |
96 | |
Ideal for hybridization and amplification |