missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Human AB Serum
Human AB serum is used in tissue engineering and cell therapy applications involving human cell types.
Brand: Corning™
This item is not returnable.
View return policy
Description
- Human AB plasma is collected from healthy donors at FDA licensed facilities located in the US
- Processed in a facility that is compliant with FDA guidelines for Class I Medical Devices located in the US
- Donor units are converted to serum and then pooled, filtered, bottled and cell culture tested for mycoplasma, endotoxin, and sterility
- Each donor unit of plasma is negative for Hepatitis B (HBsAg), Hepatitis C (anti-HCV), Human Immunodeficiency Virus-1 (HIV-1), Human Immunodeficiency Virus-2 (HIV-2), •Human Immunodeficiency Virus antibody (HIV-1Ag), and ALT by FDA approved methods.
- Proper safety precautions for potentially infectious agents must be taken despite these assurances.
- For Research Use Only
Compliance
Sera is aseptically collected and processed in a facility that is compliant with FDA guidelines for Class I Medical Devices.

Specifications
cGMP; Collected and processed according to FDA guidelines for Class I Medical Devices | |
-25° to -15°C | |
Human Serum | |
Sterile | |
≤10EU/mL | |
<20mg/dL | |
300 ±40mOsm/kg | |
6 ± 2 g/dL |
Clear, straw yellow | |
Human | |
100 mL | |
Triple-filtered, 0.1 μm | |
Used in tissue engineering and cell therapy applications involving human cell types. | |
No | |
7.8 ± 0.8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Human AB Serum > Human AB Serum; 100mL
Spot an opportunity for improvement?Share a Content Correction