Learn More
Corning™ ITS Premix Universal Culture Supplement
Stimulates proliferation of a variety of cells under serum-reduced conditions. Corning™ ITS Premix Universal Culture Supplement contains insulin, human transferrin and selenous acid, the three most universally esential components of defined culture media.
Brand: Corning™ 354350
Code : 67
Additional Details : Weight : 0.03000kg
Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
- Vialed, highly purified growth factors with high biological activity
- Broad range of highly purified natural and recombinant lymphokines
- Enriched culture supplements to be reconstituted/added in the medium of your choice
- Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
Specifications
2° to 8°C | |
Sterile-filtered | |
20 mL |
ITS Premix | |
Lyophilized from a water solution; reconstitute in 20 mL dH2O (stock solution) |