Learn More
Corning™ ITS Premix Universal Culture Supplement
Stimulates proliferation of a variety of cells under serum-reduced conditions. Corning™ ITS Premix Universal Culture Supplement contains insulin, human transferrin and selenous acid, the three most universally esential components of defined culture media.
£143.58 - £418.67
Specifications
Content And Storage | 2° to 8°C |
---|---|
Product Type | ITS Premix |
Sterility | Sterile-filtered |
Product Code | Brand | Quantity | Product Type | Formulation | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Formulation | Price | Quantity & Availability | |||
10070791
|
Corning™
354350 |
20 mL | ITS Premix | Lyophilized from a water solution; reconstitute in 20 mL dH2O (stock solution) |
£418.67
20mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10102281
|
Corning™
354351 |
5 mL | ITS Premix | Lyophilized from a water solution; reconstitute in 5 mL dH2O (stock solution) |
£143.58
5mL |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
- Vialed, highly purified growth factors with high biological activity
- Broad range of highly purified natural and recombinant lymphokines
- Enriched culture supplements to be reconstituted/added in the medium of your choice
- Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
Specifications
2° to 8°C | |
ITS Premix | |
Sterile-filtered |