Learn More
Corning™ Matrigel™ GFR Basement Membrane Matrix
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ GFR Basement Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Brand: Corning™ 354230
484.75 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Alert:
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
-20°C | |
Suitable for applications that require a highly defined basement membrane preparation. | |
Engelbreth-Holm-Swarm Mouse Tumor |
Growth factor reduced basement membrane matrix | |
10 mL |