missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PET Storage Bottles, Square with Cap
Shop All Corning Products

Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
Graduations:
100 to 1000 mL
100 to 500 mL
50 to 125 mL
50 to 250 mL
Description
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Features:
- Good for storage of media, buffers and other aqueous solutions
- Wide neck provides easier access for liquid transfer
- Screened white enamel graduations are easier to read than molded graduations
- Sterilized to SAL 10−6
- Large white marking spot for easier identification and easy opening linear tear packaging
- Individually bagged
- Validated USP Class VI, noncytotoxic and nonpyrogenic

Specifications
Specifications
Capacity | 8.45 oz., 250 mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8.45 oz. |
Color | Orange |
Diameter (English) | 2.3 in. |
Diameter (English) Inner Neck | 1.4 in. |
Diameter (Metric) Inner Neck | 37 mm |
For Use With (Application) | Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage |
Material | PET |
No. per Case | 24/Cs. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
11224009 | Certificate of Quality | 30/03/2025 | 431531 |
01524003 | Certificate of Quality | 14/12/2024 | 431531 |
23422012 | Certificate of Quality | 08/12/2023 | 431531 |
21122010 | Certificate of Quality | 03/11/2023 | 431531 |
01524003 | Certificate of Compliance | 10/04/2024 | 431531 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ PET Storage Bottles, Square with Cap >
Spot an opportunity for improvement?Share a Content Correction